• USA Home
  • 85192 - Sodium thiosulfate solution

EMAIL THIS PAGE TO A FRIEND
85192 Sigma-Aldrich

Sodium thiosulfate solution

Silver stain kit component, 0.5 % (w/v) sodium thiosulfate in H2O

Purchase

Properties

Related Categories Chemical Synthesis, Core Bioreagents, Detergents, Inorganic Salts, Research Essentials,
mol wt   158.11 g/mol
concentration   0.5 % (w/v) sodium thiosulfate in H2O
storage temp.   2-8°C
SMILES string   [Na+].[Na+].[O-]S([O-])(=O)=S
InChI   1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key   AKHNMLFCWUSKQB-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?