• USA Home
  • 85450C - Trypsin Powder, Porcine 1:250

EMAIL THIS PAGE TO A FRIEND
85450C SAFC

Trypsin Powder, Porcine 1:250

Gamma irradiated by SER-TAIN Process, dry powder

Synonym: Trypsin from porcine pancreas

Purchase

Description

Application

For trypsin digestion of peptides, use a ratio of about 1:100 to 1:20 for trypsin:peptide. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. Trypsins have also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestionsns†. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.

Trypsin Powder, Porcine 1:250 is used to release adherent cells from tissue culture plates for passaging. It was used in the isolation and culture of human endothelial cells.

Biochem/physiol Actions

Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. The optimal pH for trypsin activity is 7-9. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.

Serine protease inhibitors, including DFP, TLCK, APMSF, AEBSEF, and aprotinin, amongst others, will inhibit Trypsin.

Unit Definition

One BAEE unit will produce a ΔA253 of 0.001 per min at pH 7.6 at 25° C using BAEE as substrate. One BTEE unit = 320 ATEE units. Reaction volume = 3.2 mL (1 cm light path).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
RTECS 
YN5075000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Pipette Controller from Brand
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Enzymes & Proteins

Committed to developing and providing breakthrough problem-solving reagents and solutions, explore our comprehensive offering of enzymes and protein reagents and kits with the confidence that you wil...
Keywords: Cell signaling, Diagnostic, Enzyme-linked immunosorbent assay, Immunohistochemistry, Metabolism, Molecular biology, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

A6964 Accutase® solution, sterile-filtered, suitable for cell culture
06693 Timestrip Plus -20 °C
T6414 Trypsin inhibitor from Glycine max (soybean), solution, sterile-filtered, suitable for, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?