• USA Home
  • 855650 - (−)-N-Acetylneuraminic acid

EMAIL THIS PAGE TO A FRIEND
855650 Sigma-Aldrich

(−)-N-Acetylneuraminic acid

synthetic, crystalline

Synonym: Lactaminic acid, NANA, Sialic acid

  • CAS Number 131-48-6

  • Empirical Formula (Hill Notation) C11H19NO9

  • Molecular Weight 309.27

  •  Beilstein/REAXYS Number 1716283

  •  EC Number 205-023-1

  •  MDL number MFCD00006620

  •  PubChem Substance ID 57653055

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
form   synthetic, crystalline
optical activity   [α]24/D −32.0°, c = 1 in H2O
mp   184-186 °C (dec.) (lit.)
SMILES string   [H][C@]1(O[C@@](O)(C[C@H](O)[C@H]1NC(C)=O)C(O)=O)[C@H](O)[C@H](O)CO
InChI   1S/C11H19NO9/c1-4(14)12-7-5(15)2-11(20,10(18)19)21-9(7)8(17)6(16)3-13/h5-9,13,15-17,20H,2-3H2,1H3,(H,12,14)(H,18,19)/t5-,6+,7+,8+,9+,11-/m0/s1
InChI key   SQVRNKJHWKZAKO-PFQGKNLYSA-N

Description

Application

N-Acetylneuraminic acid (NANA, Neu5Ac) is a major component of glycoconjugates such as glycolipids, glycoproteins and proteoglycans (sialogylcoproteins) where it confers selective binding characteristics to the glycosylated component. Neu5Ac is used to study its biochemistry, metabolism and uptake in vivo and in vitro. Neu5Ac is used in the development of nanocarriers.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?