• USA Home
  • 86125 - Sulfatides from bovine brain

EMAIL THIS PAGE TO A FRIEND
86125 Sigma-Aldrich

Sulfatides from bovine brain

≥96.5% (TLC)

Synonym: Cerebroside sulfate

Purchase

Properties

assay   ≥96.5% (TLC)
solubility   chloroform/methanol (2:1): 20 mg/mL, clear, colorless to light yellow
storage temp.   −20°C

Description

Other Notes

Sales restrictions may apply

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?