• USA Home
  • 91587 - 2-Methoxy-5-(N-phthalimidinyl)benzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
91587 Sigma-Aldrich

2-Methoxy-5-(N-phthalimidinyl)benzenesulfonyl chloride

suitable for fluorescence, ≥97.0% (CHN)

Synonym: M-Phisyl chloride

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Derivatization Reagents, Derivatization Reagents HPLC,
Quality Level   100
assay   ≥97.0% (CHN)
fluorescence   λex 330 nm; λem 427 nm in chloroform (after derivatization with hexylamine)
suitability   suitable for fluorescence
SMILES string   COc1ccc(cc1S(Cl)(=O)=O)N2Cc3ccccc3C2=O
InChI   1S/C15H12ClNO4S/c1-21-13-7-6-11(8-14(13)22(16,19)20)17-9-10-4-2-3-5-12(10)15(17)18/h2-8H,9H2,1H3
InChI key   ABWFLHUIHQOEBX-UHFFFAOYSA-N

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?