• USA Home
  • 93880 - Element14® PDMS 1000

EMAIL THIS PAGE TO A FRIEND
93880 Sigma-Aldrich

Element14® PDMS 1000

viscosity ~1000 cSt 

Synonym: Dimethylpolysiloxane, Poly(dimethylsiloxane)

Purchase

Properties

vapor pressure   5 mmHg ( 20 °C)
refractive index   n20/D 1.401-1.405
viscosity   950-1050 mPa.s, neat(25 °C)
  ~1000 cSt
density   0.967-0.975 g/mL at 20 °C
SMILES string   C[Si](C)=O

Description

Legal Information

Element14 is a registered trademark of Momentive Performance Materials, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
RTECS 
JT6485000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?