• USA Home
  • 94298 - D-myo-Inositol 3,4,5-tris-phosphate tripotassium salt

EMAIL THIS PAGE TO A FRIEND
94298 Sigma-Aldrich

D-myo-Inositol 3,4,5-tris-phosphate tripotassium salt

≥85% (HPLC)

Synonym: L-myo-Inositol 1,5,6-tris-phosphate tripotassium salt

Purchase

Properties

InChI Key   NONANGHAKWRXKQ-OLYDHAFESA-K
assay   ≥85% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

Inositides have an important impact on diverse areas of cellular regulation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?