EMAIL THIS PAGE TO A FRIEND
95308 Sigma-Aldrich

L-(+)-Tartaric acid

BioUltra, ≥99.5% (T)

Synonym: (2R,3R)-(+)-Tartaric acid, L-Threaric acid

  • CAS Number 87-69-4

  • Linear Formula HO2CCH(OH)CH(OH)CO2H

  • Molecular Weight 150.09

  •  Beilstein/REAXYS Number 1725147

  •  EC Number 201-766-0

  •  MDL number MFCD00064207

  •  PubChem Substance ID 57653669

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers, Chelators More...
vapor density   5.18 (vs air)
product line   BioUltra
assay   ≥99.5% (T)
form   powder or crystals
optical activity   [α]20/D +13.5±0.5°, c = 10% in H2O
autoignition temp.   797 °F
impurities   insoluble matter, passes filter test
ign. residue   ≤0.02% (as SO4)
loss   ≤0.1% loss on drying, 110 °C
pH   1.0-2.0 (25 °C, 1 M in H2O)
mp   170-172 °C (lit.)
solubility   H2O: 1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤5 mg/kg
  phosphate (PO43-): ≤10 mg/kg
  sulfate (SO42-): ≤50 mg/kg
cation traces   Al: ≤10 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤10 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Li: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Zn: ≤5 mg/kg
λ   1 M in H2O
UV absorption   λ: 260 nm Amax: 0.3
  λ: 280 nm Amax: 0.20
SMILES string   O[C@H]([C@@H](O)C(O)=O)C(O)=O
InChI   1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1
InChI key   FEWJPZIEWOKRBE-JCYAYHJZSA-N

Description

Application

L-(+)-Tartaric acid is used in the organic synthesis of complex chiral molecules and as a reference material.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
WW7875000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?