• USA Home
  • A0255 - L-Alanine tert-butyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
A0255 Sigma-Aldrich

L-Alanine tert-butyl ester hydrochloride

  • CAS Number 13404-22-3

  • Linear Formula CH3CH(NH2)COOC(CH3)3 · HCl

  • Molecular Weight 181.66

  •  Beilstein/REAXYS Number 3562140

  •  EC Number 236-497-8

  •  MDL number MFCD00035524

  •  PubChem Substance ID 24890413

Purchase

Properties

storage temp.   −20°C
SMILES string   Cl.C[C@H](N)C(=O)OC(C)(C)C
InChI   1S/C7H15NO2.ClH/c1-5(8)6(9)10-7(2,3)4;/h5H,8H2,1-4H3;1H/t5-;/m0./s1
InChI key   WIQIWPPQGWGVHD-JEDNCBNOSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?