• USA Home
  • A0269 - 8-(6-Aminohexyl)aminoadenosine 5′-triphosphate lithium salt

EMAIL THIS PAGE TO A FRIEND
A0269 Sigma-Aldrich

8-(6-Aminohexyl)aminoadenosine 5′-triphosphate lithium salt

~90%

  • CAS Number 102029-46-9

  • Empirical Formula (Hill Notation) C16H30N7O13P3

  • Molecular Weight 621.37

  •  MDL number MFCD00056946

Purchase

Properties

assay   ~90%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?