EMAIL THIS PAGE TO A FRIEND
A0274 Sigma-Aldrich

Antimycin A2

~90%

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics by Application, Antibiotics by Mechanism of Action, Antifungal,
InChI Key   LYDAGTPXPZARPR-UHFFFAOYSA-N
assay   ~90%
Mode of action   enzyme | inhibits
antibiotic activity spectrum   fungi
storage temp.   −20°C

Description

Biochem/physiol Actions

Antimycin A2 is used to study ubiquinol (semiquinone/ubiquinone) oxidation/reduction, free radical generation, and the inhibition of cytochrome c oxidoreductase (mitochondrial complex III).

General description

Chemical structure: peptide

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?