• USA Home
  • A0275 - Antazoline methanesulfonate salt

EMAIL THIS PAGE TO A FRIEND
A0275 Supelco

Antazoline methanesulfonate salt

Synonym: 2-(N-Benzylanilinomethyl)-2-imidazoline

Purchase

Properties

SMILES string   CS(O)(=O)=O.C1CN=C(CN(Cc2ccccc2)c3ccccc3)N1

Description

Biochem/physiol Actions

Imidazoline agonist; more potent than efaroxan in inducing insulin release from β cells; H1 histamine receptor antagonist

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
RTECS 
NI6650000
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?