• USA Home
  • A0552 - (−)-N-Allylnormetazocine hydrochloride

EMAIL THIS PAGE TO A FRIEND
A0552 Supelco

(−)-N-Allylnormetazocine hydrochloride

Synonym: (−)-NANM, (−)-SKF-10047, (2α,6α)-1,2,3,4,5,6-Hexahydro-6,11-dimethyl-3-(2-propenyl)-2,6-methano-3-benzazocin-8-ol

  • CAS Number 14198-28-8

  • Empirical Formula (Hill Notation) C17H23NO · HCl

  • Molecular Weight 293.83

  •  MDL number MFCD00078553

  •  PubChem Substance ID 329770618

Purchase

Properties

InChI Key   ZTGMHFIGNYXMJV-YGCKJKTASA-N
form   neat
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?