• USA Home
  • A0686 - Anthranilyl-Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide

EMAIL THIS PAGE TO A FRIEND
A0686 Sigma-Aldrich

Anthranilyl-Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide

≥90% (HPLC)

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   BXBJVWUVNVSLAM-UHFFFAOYSA-N
assay   ≥90% (HPLC)
storage temp.   −20°C

Description

Substrates

HIV protease substrate.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?