• USA Home
  • A102 - (±)-2-Amino-4-phosphonobutyric acid

EMAIL THIS PAGE TO A FRIEND
A102 Sigma-Aldrich

(±)-2-Amino-4-phosphonobutyric acid

Synonym: (±)-AP-4

  • CAS Number 20263-07-4

  • Empirical Formula (Hill Notation) C4H10NO5P

  • Molecular Weight 183.10

  •  MDL number MFCD00013999

Purchase

Properties

SMILES string   NC(CCP(O)(O)=O)C(O)=O
InChI   1S/C4H10NO5P/c5-3(4(6)7)1-2-11(8,9)10/h3H,1-2,5H2,(H,6,7)(H2,8,9,10)
InChI key   DDOQBQRIEWHWBT-UHFFFAOYSA-N

Description

Biochem/physiol Actions

NMDA glutamate receptor antagonist.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A1910 (±)-2-Amino-4-phosphonobutyric acid, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?