• USA Home
  • A1050 - β-Asp-Phe methyl ester

EMAIL THIS PAGE TO A FRIEND
A1050 Sigma-Aldrich

β-Asp-Phe methyl ester

Synonym: N-β-L-Aspartyl-L-phenylalanine methyl ester

Purchase

Properties

mp   201-204 °C (dec.) (lit.)
storage temp.   −20°C
SMILES string   COC(=O)[C@H](Cc1ccccc1)NC(=O)C[C@H](N)C(O)=O
InChI   1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-12(17)8-10(15)13(18)19/h2-6,10-11H,7-8,15H2,1H3,(H,16,17)(H,18,19)/t10-,11-/m0/s1
InChI key   SHHIPKDJQYIJJF-QWRGUYRKSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

421677 β-Asp-Phe methyl ester, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?