• USA Home
  • A1204 - Arg-Pro p-nitroanilide acetate salt

EMAIL THIS PAGE TO A FRIEND
A1204 Sigma-Aldrich

Arg-Pro p-nitroanilide acetate salt

protease substrate

  • CAS Number 157054-08-5

  • Empirical Formula (Hill Notation) C17H25N7O4

  • Molecular Weight 391.42

  •  MDL number MFCD00274379

Purchase

Properties

storage temp.   −20°C
SMILES string   CC(O)=O.N[C@@H](CCCNC(N)=N)C(=O)N1CCC[C@H]1C(=O)Nc2ccc(cc2)[N+]([O-])=O

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Substrate for dipeptidyl aminopeptidase IV

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?