• USA Home
  • A1267 - N-(ε-Aminocaproyl)-β-D-galactopyranosylamine

EMAIL THIS PAGE TO A FRIEND
A1267 Sigma-Aldrich

N-(ε-Aminocaproyl)-β-D-galactopyranosylamine

  • CAS Number 38822-56-9

  • Empirical Formula (Hill Notation) C12H24N2O6

  • Molecular Weight 292.33

  •  MDL number MFCD00080857

Purchase

Properties

SMILES string   NCCCCCC(=O)NC1OC(CO)C(O)C(O)C1O
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?