EMAIL THIS PAGE TO A FRIEND
A1461 Sigma-Aldrich

Methyl p-anisate

≥99% (capillary GC)

Synonym: p-Anisic acid methyl ester, Methyl 4-methoxybenzoate

  • CAS Number 121-98-2

  • Linear Formula CH3OC6H4CO2CH3

  • Molecular Weight 166.17

  •  Beilstein/REAXYS Number 2208571

  •  EC Number 204-513-2

  •  MDL number MFCD00008437

  •  PubChem Substance ID 24890546

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   ≥99% (capillary GC)
bp   244-245 °C (lit.)
mp   47-51 °C (lit.)
SMILES string   COC(=O)c1ccc(OC)cc1
InChI   1S/C9H10O3/c1-11-8-5-3-7(4-6-8)9(10)12-2/h3-6H,1-2H3
InChI key   DDIZAANNODHTRB-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
RTECS 
BZ4925000

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?