• USA Home
  • A171 - 5-(N-Ethyl-N-isopropyl)amiloride

EMAIL THIS PAGE TO A FRIEND
A171 Sigma-Aldrich

5-(N-Ethyl-N-isopropyl)amiloride

Synonym: EIPA

  • CAS Number 1154-25-2

  • Empirical Formula (Hill Notation) C11H18ClN7O

  • Molecular Weight 299.76

  •  MDL number MFCD00151710

Purchase

Properties

mp   202-205 °C (lit.)
storage temp.   2-8°C
SMILES string   CCN(C(C)C)c1nc(N)c(nc1Cl)C(=O)NC(N)=N
InChI   1S/C11H18ClN7O/c1-4-19(5(2)3)9-7(12)16-6(8(13)17-9)10(20)18-11(14)15/h5H,4H2,1-3H3,(H2,13,17)(H4,14,15,18,20)
InChI key   QDERNBXNXJCIQK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A3085 5-(N-Ethyl-N-isopropyl)amiloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?