EMAIL THIS PAGE TO A FRIEND
A1791 Sigma-Aldrich

Asp-Gln

  • CAS Number 13433-13-1

  • Empirical Formula (Hill Notation) C9H15N3O6

  • Molecular Weight 261.23

  •  MDL number MFCD20122394

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?