• USA Home
  • A1915 - 2-Ethylhexyl acrylate

EMAIL THIS PAGE TO A FRIEND
A1915 SAFC

2-Ethylhexyl acrylate

Synonym: (±)-Acrylic acid 2-ethylhexyl ester

  • CAS Number 103-11-7

  • Linear Formula CH2=CHCOOCH2CH(C2H5)(CH2)3CH3

  • Molecular Weight 184.28

  •  Beilstein/REAXYS Number 1765828

  •  EC Number 203-080-7

  •  MDL number MFCD00009495

Purchase

Properties

vapor density   6.4 (vs air)
vapor pressure   0.15 mmHg ( 20 °C)
InChI Key   GOXQRTZXKQZDDN-UHFFFAOYSA-N
autoignition temp.   496 °F
expl. lim.   6.4 %
refractive index   n20/D 1.436 (lit.)
bp   215-219 °C(lit.)
density   0.885 g/mL at 25 °C (lit.)
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
1
RTECS 
AT0855000
Flash Point(F) 
174.2 °F
Flash Point(C) 
79 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

290815 2-Ethylhexyl acrylate, 98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?