EMAIL THIS PAGE TO A FRIEND
A1916 Sigma-Aldrich

Asp-Glu

≥95% (TLC)

  • CAS Number 6157-06-8

  • Empirical Formula (Hill Notation) C9H14N2O7

  • Molecular Weight 262.22

  •  MDL number MFCD00057829

  •  PubChem Substance ID 24890601

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   CKAJHWFHHFSCDT-UHFFFAOYSA-N
assay   ≥95% (TLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

L-Aspartate-L-Glutamic acid (Asp-Glu) is a precursor molecule which upon acetylation to N-acetyl-aspartyl-glutamate (NAAG) is used as a metabotropic glutamate mGlu receptor agonist.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?