EMAIL THIS PAGE TO A FRIEND
A2026 Sigma-Aldrich

Allopurinol riboside

Synonym: 1-β-D-Ribofuranosyl-1H-pyrazolo[3,4-d]pyrimidine-4-one

  • CAS Number 16220-07-8

  • Empirical Formula (Hill Notation) C10H12N4O5

  • Molecular Weight 268.23

  •  MDL number MFCD00057096

  •  PubChem Substance ID 24890617

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   KFQUAMTWOJHPEJ-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Biochem/physiol Actions

An anti-leishmanial agent

Application

Allopurinol riboside is an experimental agent for the treatment of leishmaniasis and American trypanosomiasis. Allopurinol riboside is an inhibitor of orotidine 5′-monophosphate (OMP) decarboxylase from Plasmodium falciparum ((PfODCase, EC 4.1.1.23).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?