• USA Home
  • A2283 - Angiotensinogen from porcine plasma

EMAIL THIS PAGE TO A FRIEND
A2283 Sigma-Aldrich

Angiotensinogen from porcine plasma

lyophilized powder, 1,500-3,000 units/g protein

Synonym: Angiotensinogen from hog plasma, Hypertensinogen, Renin substrate

Purchase

Properties

Related Categories Angiotensins, Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology More...
form   lyophilized powder
specific activity   1,500-3,000 units/g protein
composition   Protein, ~65% modified Warburg-Christian
application(s)   ligand binding assay: suitable
color   white
suitability   suitable for measuring renin by RIA
storage temp.   −20°C
SMILES string   CCC(C)C(NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CC(O)=O)C(C)C)C(=O)NC(Cc2cnc[nH]2)C(=O)N3CCCC3C(=O)NC(Cc4ccccc4)C(=O)NC(Cc5cnc[nH]5)C(=O)NC(CC(C)C)C(=O)NC(CC(C)C)C(=O)NC(C(C)C)C(=O)NC(Cc6ccc(O)cc6)C(=O)NC(CO)C(O)=O
InChI   1S/C85H123N21O20/c1-11-48(10)70(105-78(119)62(35-51-23-27-55(109)28-24-51)100-80(121)68(46(6)7)103-72(113)57(19-15-29-91-85(87)88)94-71(112)56(86)38-67(110)111)82(123)101-64(37-53-40-90-43-93-53)83(124)106-30-16-20-66(106)79(120)98-60(33-49-17-13-12-14-18-49)74(115)97-63(36-52-39-89-42-92-52)76(117)95-58(31-44(2)3)73(114)96-59(32-45(4)5)77(118)104-69(47(8)9)81(122)99-61(34-50-21-25-54(108)26-22-50)75(116)102-65(41-107)84(125)126/h12-14,17-18,21-28,39-40,42-48,56-66,68-70,107-109H,11,15-16,19-20,29-38,41,86H2,1-10H3,(H,89,92)(H,90,93)(H,94,112)(H,95,117)(H,96,114)(H,97,115)(H,98,120)(H,99,122)(H,100,121)(H,101,123)(H,102,116)(H,103,113)(H,104,118)(H,105,119)(H,110,111)(H,125,126)(H4,87,88,91)
InChI key   XJFQCYIFOWHHFN-UHFFFAOYSA-N

Description

Unit Definition

One unit will yield 1.0 nmole of angiotensinI in the presence of renin at pH 6.0 at 37 °C measured by RIA.

Physical form

Lyophilized powder containing sodium citrate.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?