• USA Home
  • A2291 - N-Acetyl-Ala-Ala-Pro-Ala β-naphthylamide

EMAIL THIS PAGE TO A FRIEND
A2291 Sigma-Aldrich

N-Acetyl-Ala-Ala-Pro-Ala β-naphthylamide

  • CAS Number 102029-65-2

  • Empirical Formula (Hill Notation) C26H33N5O5

  • Molecular Weight 495.57

Purchase

Properties

storage temp.   −20°C

Description

Substrates

Chromogenic substrate for proteases.

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?