• USA Home
  • A230 - 4-Amino-5-hexynoic acid

EMAIL THIS PAGE TO A FRIEND
A230 Sigma-Aldrich

4-Amino-5-hexynoic acid

solid

Synonym: γ-Acetylenic GABA

  • Empirical Formula (Hill Notation) C6H9NO2

  • Molecular Weight 127.14

  •  MDL number MFCD00468056

  •  PubChem Substance ID 24278145

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Enzyme Inhibitors, GABA-Transaminase, Neuroscience,
InChI Key   BJNIHWSOVCDBHS-UHFFFAOYSA-N
form   solid
color   white
solubility   H2O: 5 mg/mL
Gene Information   human ... ABAT(18)

Description

Biochem/physiol Actions

Inhibitor of GABA transaminase.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MR0180450

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?