EMAIL THIS PAGE TO A FRIEND
A2343 Sigma-Aldrich

L-Ascorbic acid

meets USP testing specifications

Synonym: L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

  • CAS Number 50-81-7

  • Empirical Formula (Hill Notation) C6H8O6

  • Molecular Weight 176.12

  •  Beilstein/REAXYS Number 84272

  •  EC Number 200-066-2

  •  MDL number MFCD00064328

  •  PubChem Substance ID 24890649

Purchase

Properties

Related Categories Analytical/Chromatography, Biochemicals and Reagents, Core Bioreagents, Pharmacopoeia, Pharmacopoeia A-Z,
biological source   synthetic
InChI Key   CIWBSHSKHKDKBQ-JLAZNSOCSA-N
assay   99.0-100.5%
form   solid
pH   1.0-2.5 (25 °C, 176 g/L in water)
mp   190-194 °C (dec.)
solubility   water: soluble 176 g/L at 20 °C
suitability   meets USP testing specifications
Featured Industry   Pharmaceutical (small molecule)
Gene Information   human ... SLC23A2(9962)

Description

Biochem/physiol Actions

Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid functions as an enzymatic cofactor for multiple enzymes, serving as an electron donor for monooxygenases and dioxygenases. Ascorbic acid also functions as a powerful antioxidant, particularly in regards to reactive oxygen species.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CI7650000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?