• USA Home
  • A2345 - Arachidonylcyclopropylamide

EMAIL THIS PAGE TO A FRIEND
A2345 Sigma

Arachidonylcyclopropylamide

≥97%

Synonym: ACPA

  • Empirical Formula (Hill Notation) C23H37NO

  • Molecular Weight 343.55

  •  MDL number MFCD02683582

Purchase

Properties

assay   ≥97%
solubility   DMSO: soluble
  H2O: insoluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Potent and selective neuronal CB1 cannabinoid receptor agonist.

Caution

Photosensitive; store under argon or nitrogen.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?