EMAIL THIS PAGE TO A FRIEND
A2416 Sigma-Aldrich

Arg-Ser-Arg

  • CAS Number 115035-42-2

  • Empirical Formula (Hill Notation) C15H31N9O5

  • Molecular Weight 417.46

  •  MDL number MFCD00058510

Purchase

Properties

storage temp.   −20°C
SMILES string   NC(CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(O)=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?