• USA Home
  • A2656 - Adenosine 5′-succinate

EMAIL THIS PAGE TO A FRIEND
A2656 Sigma

Adenosine 5′-succinate

  • CAS Number 102029-71-0

  • Empirical Formula (Hill Notation) C14H17N5O7

  • Molecular Weight 367.31

  •  MDL number MFCD00037996

Purchase

Properties

storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?