• USA Home
  • A2677 - Atrial Natriuretic Peptide fragment 4-28 rat

EMAIL THIS PAGE TO A FRIEND
A2677 Sigma-Aldrich

Atrial Natriuretic Peptide fragment 4-28 rat

≥97% (HPLC)

Synonym: Auriculin® B, rANF 4-28

  • CAS Number 90052-57-6

  • Empirical Formula (Hill Notation) C113H177N39O35S2

  • Molecular Weight 2705.98

  •  MDL number MFCD00076239

Purchase

Properties

assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Nppa(24602)

Description

Biochem/physiol Actions

Atrial natriuretic peptide (ANP) is synthesized in myoendocrine cells of the heart from which it is released into the circulation. It exerts natriuretic, diuretic, and vasodilatory effects through stimulation of guanylate cyclase-linked NPR-A receptors. It plays an important role in blood volume and blood pressure homeostasis.

Legal Information

Auriculin is a registered trademark of California Biotechnology, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?