• USA Home
  • A2766 - 6-Aminopyridine-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
A2766 Sigma-Aldrich

6-Aminopyridine-3-carboxylic acid

Synonym: 6-Aminonicotinic acid

  • CAS Number 3167-49-5

  • Empirical Formula (Hill Notation) C6H6N2O2

  • Molecular Weight 138.12

  •  Beilstein/REAXYS Number 115992

  •  EC Number 221-630-4

  •  MDL number MFCD00006326

Purchase

Properties

mp   >300 °C (lit.)
SMILES string   Nc1ccc(cn1)C(O)=O
InChI   1S/C6H6N2O2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H2,7,8)(H,9,10)
InChI key   ZCIFWRHIEBXBOY-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

216879 6-Aminopyridine-3-carboxylic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?