EMAIL THIS PAGE TO A FRIEND
A3102 Sigma-Aldrich

Acetic acid

meets EP, BP, USP testing specifications

Synonym: Glacial acetic acid

  • CAS Number 64-19-7

  • Linear Formula CH3CO2H

  • Molecular Weight 60.05

  •  Beilstein/REAXYS Number 506007

  •  EC Number 200-580-7

  •  MDL number MFCD00036152

Purchase

Properties

vapor density   2.07 (vs air)
autoignition temp.   800 °F
expl. lim.   16 %, 92 °F
  4 %, 59 °F
refractive index   n20/D 1.371 (lit.)
bp   117-118 °C (lit.)
mp   16.2 °C (lit.)
density   1.049 g/mL at 25 °C (lit.)
suitability   meets EP, BP, USP testing specifications
SMILES string   CC(O)=O
InChI   1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)
InChI key   QTBSBXVTEAMEQO-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2789PSN1 3(8) / PGII
WGK Germany 
1
RTECS 
AF1225000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?