• USA Home
  • A3146 - 2-Amino-4,6-dichloropyrimidine

EMAIL THIS PAGE TO A FRIEND
A3146 Sigma-Aldrich

2-Amino-4,6-dichloropyrimidine

~95%

  • CAS Number 56-05-3

  • Empirical Formula (Hill Notation) C4H3Cl2N3

  • Molecular Weight 163.99

  •  Beilstein/REAXYS Number 118454

  •  EC Number 200-253-9

  •  MDL number MFCD00006090

Purchase

Properties

assay   ~95%
mp   219-222 °C (lit.)
SMILES string   Nc1nc(Cl)cc(Cl)n1
InChI   1S/C4H3Cl2N3/c5-2-1-3(6)9-4(7)8-2/h1H,(H2,7,8,9)
InChI key   JPZOAVGMSDSWSW-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UV6260485

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A48601 2-Amino-4,6-dichloropyrimidine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?