• USA Home
  • A3390 - 3′-O-Acetyl-2′-deoxycytidine

EMAIL THIS PAGE TO A FRIEND
A3390 Sigma-Aldrich

3′-O-Acetyl-2′-deoxycytidine

~98%

  • CAS Number 72560-69-1

  • Empirical Formula (Hill Notation) C11H15N3O5

  • Molecular Weight 269.25

  •  MDL number MFCD00057458

  •  PubChem Substance ID 24890764

Purchase

Properties

assay   ~98%
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?