EMAIL THIS PAGE TO A FRIEND
A3396 Sigma-Aldrich

Acetonitrile

≥99.5%

Synonym: ACN, Methyl cyanide

  • CAS Number 75-05-8

  • Linear Formula CH3CN

  • Molecular Weight 41.05

  •  Beilstein/REAXYS Number 741857

  •  EC Number 200-835-2

  •  MDL number MFCD00001878

  •  eCl@ss 39031501

  •  PubChem Substance ID 329770720

Purchase

Properties

vapor density   1.41 (vs air)
vapor pressure   72.8 mmHg ( 20 °C)
InChI Key   WEVYAHXRMPXWCK-UHFFFAOYSA-N
assay   ≥99.5%
form   liquid
autoignition temp.   973 °F
expl. lim.   16 %
color   colorless
refractive index   n20/D 1.344 (lit.)
bp   81-82 °C (lit.)
mp   −48 °C (lit.)
solubility   water: miscible
density   0.786 g/mL at 25 °C (lit.)
suitability   suitable (Maldi-TOF mass spectroscopy)

Description

General description

Acetonitrile (MeCN), an aliphatic nitrile, is a colorless liquid with a pleasant odor. It is widely used as a solvent and intermediate in organic syntheses. It is transparent to UV-visible light, making it highly applicable in spectrophotometric and fluorimetric techniques. As MeCN has low viscosity, high elution strength and is highly miscible in water, it is utilized as a mobile phase component in many chromatographic techniques. It also plays a major role as an extractant medium in liquid-liquid extraction, solid-phase extraction or microextraction. Its infrared spectrum has been recorded. Synthesis of alkyl hydroperoxides in MeCN by alkane oxidation with hydrogen peroxide in the presence of iron complexes has been studied. The hydrogenation of MeCN to form ethylamine using Co-B amorphous alloy catalyst has been investigated.

Application

Acetonitrile (MeCN) was used to constitute mobile phase A, which is employed in the UPLC (ultra-performance liquid chromatography) based separation of underivatized amino acids derived from monoclonal antibody producing Chinese hamster ovary (CHO) cells.
It may be used as solvent in the following processes:
• Determination of pKa values of organic superbases using isodensity polarization continuum model (IPCM).
• Preparation of 1,2-azidoalcohols and 1,2-azidoamines via cerium(III) chloride assisted ring opening of epoxides and aziridines by sodium azide.
• Synthesis of cyano-bearing indolinones by oxidative arylalkylation of olefins in the presence of palladium catalyst.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1648 3 / PGII
WGK Germany 
2
RTECS 
AL7700000
Flash Point(F) 
35.6 °F
Flash Point(C) 
2 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?