• USA Home
  • A3505 - Sodium 4-aminosalicylate dihydrate

EMAIL THIS PAGE TO A FRIEND
A3505 Sigma-Aldrich

Sodium 4-aminosalicylate dihydrate

Synonym: 4-Amino-2-hydroxybenzoic acid sodium salt, 4-Amino-salicylic acid sodium salt

Purchase

Properties

InChI Key   GMUQJDAYXZXBOT-UHFFFAOYSA-M
form   crystalline
color   white
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?