EMAIL THIS PAGE TO A FRIEND
A3526 Sigma-Aldrich

Asp-Ser-Asp-Pro-Arg

≥90% (HPLC)

Synonym: Hamburger pentapeptide, IgE peptide III

  • CAS Number 62087-72-3

  • Empirical Formula (Hill Notation) C22H36N8O11

  • Molecular Weight 588.57

  •  MDL number MFCD00081022

Purchase

Properties

assay   ≥90% (HPLC)
storage temp.   −20°C
SMILES string   N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(O)=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CCCNC(N)=N)C(O)=O
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?