• USA Home
  • A3582 - 3-Acetylumbelliferyl β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
A3582 Sigma-Aldrich

3-Acetylumbelliferyl β-D-glucopyranoside

≥95%

Synonym: 3-Acetyl-7-coumarinyl-β-D-glucopyranoside, 3-Acetyl-7-(β-D-glucopyranosyloxy)coumarin

  • CAS Number 20943-16-2

  • Empirical Formula (Hill Notation) C17H18O9

  • Molecular Weight 366.32

  •  Beilstein/REAXYS Number 1268871

  •  MDL number MFCD00037378

  •  PubChem Substance ID 24890787

Purchase

Description

Substrates

Fluorogenic substrate for β-glucosidase.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Fermentation: Enzymes, Substrates and SCFA

The influence of probiotics on the metabolic activity of intestinal microbiota may be evaulated by detecting bacterial enzymatic activity in fecal samples, specifically β-glucosidase, β-glucuronidase...
Chloe McClanahan
Biofiles Edition 6.2
Keywords: Absorption, Cancer, Carcinogens, Cell proliferation, Chromatography, Enzyme activity, Gas chromatography, Gastrointestinal, High performance liquid chromatography, Metabolites, Prebiotics, Probiotics, Thin layer chromatography

Probiotics and Human Health

At the beginning of the 20th century, probiotics were thought to have a beneficial effect to the host by improving the intestinal microbial balance. Through his studies and observations, the Russian ...
Chloe McClanahan
BioFiles Edition 6.2
Keywords: AGE, Absorption, Adhesion, Amplification, Anaerobic, Antimicrobials, Cancer, Carcinogens, Cell proliferation, Chromatography, Coagulation, Detoxification, Diagnostic, Diseases, Enzyme activity, Fermentation, Gas chromatography, Gastrointestinal, High performance liquid chromatography, Inflammation, Magnetic resonance spectroscopy, Metabolism, Metabolites, Molecular biology, Nucleic acid hybridization, PAGE, Pharmaceutical, Phosphorylations, Polymerase chain reaction, Prebiotics, Probiotics, Reductions, Respiratory, Sequencing, Thin layer chromatography, Titrations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?