• USA Home
  • A3690 - L-(+)-2-Amino-3-phosphonopropionic acid

EMAIL THIS PAGE TO A FRIEND
A3690 Sigma

L-(+)-2-Amino-3-phosphonopropionic acid

≥98%

Synonym: (R)-(+)-2-Amino-3-phosphonopropionic acid, 3-Phosphono-L-alanine, L-AP3

  • CAS Number 23052-80-4

  • Empirical Formula (Hill Notation) C3H8NO5P

  • Molecular Weight 169.07

  •  Beilstein Registry Number 3649605

  •  MDL number MFCD00083169

Purchase

Properties

InChI Key   LBTABPSJONFLPO-REOHCLBHSA-N
assay   ≥98%

Description

Biochem/physiol Actions

Potent antagonist at metabotropic glutamic acid receptors.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?