EMAIL THIS PAGE TO A FRIEND
A3711 Sigma-Aldrich

AA-861

≥98% (HPLC)

Synonym: 2-(12-Hydroxydodeca-5,10-diynyl)-3,5,6-trimethyl-p-benzoquinone

Purchase

Properties

assay   ≥98% (HPLC)
solubility   ethanol: soluble
storage temp.   2-8°C
Gene Information   human ... ALOX5(240)
rat ... Alox5(25290)

Description

Biochem/physiol Actions

5-Lipoxygenase inhibitor.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

The Role of Inflammation in Neurodegeneration

It is well established that the brain can sense and react to peripheral insults through the rapid induction of fever and other neuroendocrine changes in response to events such as infection and tissu...
Scott Hauser, Technology Transfer Specialist, Sigma® Life Science
Biofiles, Vol. 8, No. 19
Keywords: Adhesion, Central Nervous System, Diseases, Gene expression, Inflammation, Neurodegenerative Diseases, Reductions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?