• USA Home
  • A3719 - S-(5′-Adenosyl)-L-methionine-(methyl-3H)

EMAIL THIS PAGE TO A FRIEND
A3719 Sigma-Aldrich

S-(5′-Adenosyl)-L-methionine-(methyl-3H)

≥95% (HPLC), buffered aqueous solution

Synonym: SAM-(methyl-3H)

  • CAS Number 111093-45-9

  • Empirical Formula (Hill Notation) C15H23N6O5S

  • Molecular Weight 399.45

  •  MDL number MFCD00135929

Purchase

Properties

assay   ≥95% (HPLC)
form   buffered aqueous solution
extent of labeling   5-15 Ci/mmol
shipped in   dry ice
storage temp.   −20°C

Description

Physical form

Hydrochloric acid solution, pH 2-2.5, containing 10% ethanol

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
1
Flash Point(F) 
136 °F
Flash Point(C) 
58 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?