• USA Home
  • A3789 - 4-Aminophenyl β-D-N,N′-diacetylchitobioside

EMAIL THIS PAGE TO A FRIEND
A3789 Sigma-Aldrich

4-Aminophenyl β-D-N,N′-diacetylchitobioside

~95%

  • CAS Number 102029-83-4

  • Empirical Formula (Hill Notation) C22H33N3O11

  • Molecular Weight 515.51

Purchase

Properties

assay   ~95%
storage temp.   2-8°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?