EMAIL THIS PAGE TO A FRIEND
A3885 Sigma-Aldrich

Anthracene

≥99%, powder

Synonym: Anthraxcene, Paranaphthalene

  • CAS Number 120-12-7

  • Empirical Formula (Hill Notation) C14H10

  • Molecular Weight 178.23

  •  Colour Index Number 10790

  •  Beilstein/REAXYS Number 1905429

  •  EC Number 204-371-1

  •  MDL number MFCD00001240

Purchase

Properties

vapor density   6.15 (vs air)
vapor pressure   1 mmHg ( 145 °C)
InChI Key   MWPLVEDNUUSJAV-UHFFFAOYSA-N
assay   ≥99%
form   powder
autoignition temp.   1004 °F
color   white
bp   340 °C(lit.)
mp   210-215 °C (lit.)
solubility   alcohols: soluble
  benzene: soluble
  chloroform: soluble
  hydronaphthalenes: soluble
  supercritical carbon dioxide: soluble

Description

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Safety & Documentation

Safety Information

RIDADR 
UN 3077 9/PG 3
WGK Germany 
2
RTECS 
CA9350000
Flash Point(F) 
250 °F
Flash Point(C) 
121 °C

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

141062 Anthracene, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?