• USA Home
  • A4009 - Adenylyl(2′→5′)cytidine ammonium salt

EMAIL THIS PAGE TO A FRIEND
A4009 Sigma-Aldrich

Adenylyl(2′→5′)cytidine ammonium salt

  • CAS Number 102029-84-5

  • Empirical Formula (Hill Notation) C19H25N8O11P

  • Molecular Weight 572.42

  •  MDL number MFCD00078864

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?