• USA Home
  • A4032 - β-Ala-Trp-Met-Asp-Phe amide acetate salt

EMAIL THIS PAGE TO A FRIEND
A4032 Sigma

β-Ala-Trp-Met-Asp-Phe amide acetate salt

≥90% (HPLC)

Synonym: des-Boc-Pentagastrin

  • CAS Number 21880-05-7

  • Empirical Formula (Hill Notation) C32H41N7O7S

  • Molecular Weight 667.78

Purchase

Properties

assay   ≥90% (HPLC)
storage temp.   −20°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?