• USA Home
  • A4126 - N-Acetyl-L-phenylalanine

EMAIL THIS PAGE TO A FRIEND
A4126 Sigma

N-Acetyl-L-phenylalanine

Sigma Grade

Purchase

Properties

grade   Sigma Grade
InChI Key   CBQJSKKFNMDLON-JTQLQIEISA-N
mp   171-173 °C (lit.)
storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

857459 N-Acetyl-L-phenylalanine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?