• USA Home
  • A4170 - 2-Acetamido-2-deoxy-1-thio-β-D-glucopyranose 3,4,6-triacetate

EMAIL THIS PAGE TO A FRIEND
A4170 Sigma

2-Acetamido-2-deoxy-1-thio-β-D-glucopyranose 3,4,6-triacetate

  • CAS Number 51450-09-0

  • Empirical Formula (Hill Notation) C14H21NO8S

  • Molecular Weight 363.38

  •  MDL number MFCD00012355

Purchase

Properties

storage temp.   −20°C
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?