• USA Home
  • A4172 - 1,6-Anhydro-β-D-glucose 2,3,4-tribenzoate

EMAIL THIS PAGE TO A FRIEND
A4172 Sigma-Aldrich

1,6-Anhydro-β-D-glucose 2,3,4-tribenzoate

  • CAS Number 23567-05-7

  • Empirical Formula (Hill Notation) C27H22O8

  • Molecular Weight 474.46

Purchase
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?